Learn More
Perkin Elmer US LLC Deuterium Lamp (PDA) for Flexar/Series 200/200a EP
Catalog No. 502113166
Encompass_Preferred
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium, tungsten or xenon source provides outstanding ultraviolet and true visible performance. Exceptional performance anywhere in the detectors 190 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.
Catalog No. 50-211-3166 Supplier Perkin Elmer US LLC Supplier No. N2925030
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium, tungsten or xenon source provides outstanding ultraviolet and true visible performance. Exceptional performance anywhere in the detectors 190 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.