Learn More
Perkin Elmer US LLC UV/Vis Deuterium Detector Lamp
Catalog No. 509044156
Encompass_Preferred
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium source provides outstanding ultraviolet and true visible performance anywhere in the detector’s 190 – 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.
Catalog No. 50-904-4156 Supplier Perkin Elmer US LLC Supplier No. N2920149
Our extensive quality control and inspection process demands the very best quality sources. Choosing a PerkinElmer deuterium source provides outstanding ultraviolet and true visible performance anywhere in the detector’s 190 – 700 nm wavelength range. Lamp changes are quick and easy due to a unique self-aligning lamp mount.