Learn More
Agilent Technologies Long-Life Deuterium VWD Lamp, RFID Tag, 1 Ea
Catalog No. NC2212302
Encompass
The InfinityLab long-life deuterium lamp is designed for optimal HPLC/UV analysis, delivering high signal intensity, minimal baseline drift, and enhanced signal-to-noise ratios. Engineered specifically for Agilent variable wavelength detectors, this lamp ensures precise alignment and performance, directly contributing to improved detection limits and consistent analytical results. Each unit is equipped with an RFID tag for automatic detector integration, maintenance tracking, and simplified lamp counter resets.
  • Greater than 2,000 hr operating life
  • Higher initial intensity across 190 nm to 330 nm range
  • Reduced stabilization time following ignition
  • Bulky cap design for UV light emission safety
  • Double-insulated cable for operator protection
  • Rigorous testing for alignment, intensity, and operating voltage
  • Optimized lamp shape for precise detector interface
This item is not returnable. View return policy
Catalog No. NC2212302 Supplier Agilent Technologies Supplier No. G131460101
The InfinityLab long-life deuterium lamp is designed for optimal HPLC/UV analysis, delivering high signal intensity, minimal baseline drift, and enhanced signal-to-noise ratios. Engineered specifically for Agilent variable wavelength detectors, this lamp ensures precise alignment and performance, directly contributing to improved detection limits and consistent analytical results. Each unit is equipped with an RFID tag for automatic detector integration, maintenance tracking, and simplified lamp counter resets.
  • Greater than 2,000 hr operating life
  • Higher initial intensity across 190 nm to 330 nm range
  • Reduced stabilization time following ignition
  • Bulky cap design for UV light emission safety
  • Double-insulated cable for operator protection
  • Rigorous testing for alignment, intensity, and operating voltage
  • Optimized lamp shape for precise detector interface