Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium oxide, 99.95+% (Isotopic)

Catalog No. AA43700SA
Change view
Click to view available options
Quantity:
10 Pk.
2 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43700SA 2 Pk.
AA43700KV 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43700SA Supplier Thermo Scientific Chemicals Supplier No. 043700SA

missing translation for 'validMainframeMsg'

CAS: 7789-20-0 | H2O | 20.03 g/mol

Deuterium oxide is used in nuclear magnetic resonance spectroscopy (NMR). It is also useful in the identification of labile hydrogens. As a source of deuterium, it is utilized for preparing specifically labeled isotopologs of organic compounds. It is often used instead of water for analyzing proteins in solution by using fourier transform infrared spectroscopy (FTIR). It finds application in certain types of nuclear reactors and in tritium production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-20-0
Molecular Formula H2O
Molecular Weight (g/mol) 20.03
MDL Number MFCD00044636
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
PubChem CID 24602
ChEBI CHEBI:41981
IUPAC Name [(²H)oxy](²H)
SMILES [2H]O[2H]

Specifications

Physical Form Liquid
Color Colorless
Sensitivity Moisture
Refractive Index 1.338
Density 1.105
Quantity 2 Pk.
Vapor Pressure 5mm Hg at 25°C
Odor Odorless
Melting Point 3.82°C
Boiling Point 101.4°C
Assay Percent Range >99.95%
Solubility Information Miscible with water.
Formula Weight 20.03
Chemical Name or Material Deuterium oxide
Show More Show Less
EINECSNumber 232-148-9
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.