Learn More
Sigma Aldrich Fine Chemicals Biosciences Deuterium oxide | 7789-20-0 | MFCD00044636 | 100mL
Catalog No. NC1031189
Encompass
Deuterium oxide | 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt | MW:20.03 | 7789-20-0 | MFCD00044636 | 100mL
Catalog No. NC1031189 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 450510100ML
Deuterium oxide | 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt | MW:20.03 | 7789-20-0 | MFCD00044636 | 100mL