Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium oxide, for NMR, 99.8 atom % D

Catalog No. AC166301000
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166301000 100 mL
AC166300100 10 mL
AC166300250 25 mL
AC166305000 500 mL
AC166300010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC166301000 Supplier Thermo Scientific Chemicals Supplier No. 166301000
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7789-20-0 | H2O | 20.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-20-0
Molecular Formula H2O
Molecular Weight (g/mol) 20.03
MDL Number MFCD00044636
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
PubChem CID 24602
ChEBI CHEBI:41981
IUPAC Name [(²H)oxy](²H)
SMILES [2H]O[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 100 mL
Density 1.1050g/mL
Specific Gravity 1.105
Refractive Index 1.328
Viscosity 1.25 mPa.s (20°C)
Deuterium (D) 99.75% min.
Melting Point 3.8°C
Boiling Point 101.4°C
Linear Formula D2O
Merck Index 15, 2942
Solubility Information Solubility in water: miscible, exothermic reaction.
Formula Weight 20.02
Grade NMR
Chemical Name or Material Deuterium oxide, 99.8 atom % D
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 232-148-9
RTECSNumber ZC0230000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.