Learn More
Cayman Chemical Deuterated Arachidonic Acid CYP450 Metabolite MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502982909
Encompass_Preferred
Contains deuterated forms of HETE and EET metabolites that have been shown to be involved in either vasoconstriction or hypertension; designed for direct use in LC-MS applications; each ampule contains at least 1 ml
Catalog No. 50-298-2909 Supplier Cayman Chemical Supplier No. 206671 EA
Contains deuterated forms of HETE and EET metabolites that have been shown to be involved in either vasoconstriction or hypertension; designed for direct use in LC-MS applications; each ampule contains at least 1 ml