Learn More
Cayman Chemical DeuTRated LInoleIc ACD Oxyl
SDP
Catalog No. 502980684
Encompass_Preferred
Contains deuterated forms of (±)12(13)-DiHOME, 13(S)-HODE, 13-OxoODE, and (±)12(13)-EpOME, EAch of which have been shown to be involved in the physiological or pathological responses related to cell proliferation or oxidative burst and are potential markers for (or potential contributors to) various human diseases
Catalog No. 50-298-0684 Supplier Cayman Chemical Supplier No. 207951 EA
Add to Cart
Add to Cart
Contains deuterated forms of (±)12(13)-DiHOME, 13(S)-HODE, 13-OxoODE, and (±)12(13)-EpOME, EAch of which have been shown to be involved in the physiological or pathological responses related to cell proliferation or oxidative burst and are potential markers for (or potential contributors to) various human diseases