Learn More
Miles Scientific DEV CHAMBER FOR 20X20 PLATES
SDP
Catalog No. NC9996363
Encompass_Preferred
Glass Rectangular Developing Chamber for 20x20cm plates, Sturdy, molded-glass block of clear glass with flat surface walls, ground lip of uniform flatness, Beveled inside and outside to remove any sharp edges.Comes with flat, clear glass li
Catalog No. NC9996363 Supplier Miles Scientific Supplier No. A7022
Glass Rectangular Developing Chamber for 20x20cm plates, Sturdy, molded-glass block of clear glass with flat surface walls, ground lip of uniform flatness, Beveled inside and outside to remove any sharp edges.Comes with flat, clear glass li