Learn More
Wilmad Labglass DEV TANK TLC 25X75 95MM
Catalog No. 50803080
Encompass
Developing Tank, TLC, Cylindrical, Screw Cap. Plate Width x Length: 25mm x 75mm; Approximate Height: 95mm. Glass cylindrical developing chambers w/ Flat bottom and molded plastic caps.
Catalog No. 50-803-080 Supplier Wilmad Labglass Supplier No. LG4597098
Developing Tank, TLC, Cylindrical, Screw Cap. Plate Width x Length: 25mm x 75mm; Approximate Height: 95mm. Glass cylindrical developing chambers w/ Flat bottom and molded plastic caps.