Learn More
Wilmad Labglass DEV TANK TLC 50X100 120MM
Catalog No. 50803314
Encompass
Developing Tank, TLC, Cylindrical, Screw Cap. Plate Width x Length: 50mm x 100mm; Approximate Height: 120mm. Glass cylindrical developing chambers w/ Flat bottom and molded plastic caps.
Catalog No. 50-803-314 Supplier Wilmad Labglass Supplier No. LG4597100
Developing Tank, TLC, Cylindrical, Screw Cap. Plate Width x Length: 50mm x 100mm; Approximate Height: 120mm. Glass cylindrical developing chambers w/ Flat bottom and molded plastic caps.