Learn More
Medchemexpress LLC Devaleryl valsartan impurity | 676129-92-3 | MFCD30342453 | 100.0% | 351.40 g/mol | C19H21N5O2 | 5 MG
SDP
Catalog No. 5000262259
Encompass_Preferred
Devaleryl valsartan impurity is an intermediate and analytical reference standard used in the synthesis and impurity profiling of the antihypertensive drug valsartan. It is supplied as a high-purity research reagent for analytical method development, impurity identification, and synthetic verification, and is not intended for human or clinical use.
  • High purity (99.98%) suitable for reference-standard applications.
  • Molecular weight 351.40 g/mol and formula C19H21N5O2 for precise characterization.
  • Available in small pack sizes appropriate for method development and reference work.
  • Useful as a synthetic intermediate for route verification and impurity studies.
  • Provided with supporting documentation, including COA and SDS, to support quality control.
Catalog No. 50-002-62259 Supplier Medchemexpress LLC Supplier No. HY1312805MG
Devaleryl valsartan impurity is an intermediate and analytical reference standard used in the synthesis and impurity profiling of the antihypertensive drug valsartan. It is supplied as a high-purity research reagent for analytical method development, impurity identification, and synthetic verification, and is not intended for human or clinical use.
  • High purity (99.98%) suitable for reference-standard applications.
  • Molecular weight 351.40 g/mol and formula C19H21N5O2 for precise characterization.
  • Available in small pack sizes appropriate for method development and reference work.
  • Useful as a synthetic intermediate for route verification and impurity studies.
  • Provided with supporting documentation, including COA and SDS, to support quality control.