Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Devarda's Alloy, For Analysis, EMSURE™ MilliporeSigma™

Catalog No. M1053410250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1053410250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1053410250 Supplier MilliporeSigma Supplier No. 1.05341.0250
Add to Cart
Add to Cart

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8049-11-4
Molecular Formula Al9Cu10Zn
Molecular Weight (g/mol) 943.67
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
Synonym devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
PubChem CID 16211762
IUPAC Name aluminum;copper;zinc
SMILES [Al].[Cu].[Zn]

Specifications

Color Gray
Melting Point 490°C to 560°C
Boiling Point 906°C
Density 5.79 g/3 (20 °C)
Quantity 250 g
Packaging Plastic Bottle
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.