Learn More
Medchemexpress LLC Devazepide | 103420-77-5 | MFCD00864500 | >98.0% | 408.45 | C25H20N4O2 | 25 MG
SDP
Catalog No. 5000243637
Encompass_Preferred
Devazepide is an analytical reference standard of a potent, selective cholecystokinin (CCK1) receptor antagonist used in pharmacological research and analytical method development. It is provided for qualitative and quantitative assays and instrument calibration. CAS 103420-77-5; formula C25H20N4O2; MW 408.45.
  • Analytical reference standard for method development and calibration.
  • Suitable for HPLC, GC, and MS applications.
  • High purity >98.0% (HPLC).
  • Molecular formula C25H20N4O2; molecular weight 408.45.
  • Available in small pack sizes for analytical workflows (25 mg variant).
  • Certificate of Analysis provides assay and storage instructions.
Catalog No. 50-002-43637 Supplier Medchemexpress LLC Supplier No. HY10630125MG
Devazepide is an analytical reference standard of a potent, selective cholecystokinin (CCK1) receptor antagonist used in pharmacological research and analytical method development. It is provided for qualitative and quantitative assays and instrument calibration. CAS 103420-77-5; formula C25H20N4O2; MW 408.45.
  • Analytical reference standard for method development and calibration.
  • Suitable for HPLC, GC, and MS applications.
  • High purity >98.0% (HPLC).
  • Molecular formula C25H20N4O2; molecular weight 408.45.
  • Available in small pack sizes for analytical workflows (25 mg variant).
  • Certificate of Analysis provides assay and storage instructions.