Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Devazepide | 103420-77-5 | MFCD00864500 | ≥98.0% | 408.45 | C25H20N4O2 | 50 MG
SDP

Catalog No. 5000243395
Encompass_Preferred

Devazepide is an analytical reference standard of a selective cholecystokinin (CCK) receptor antagonist intended for research and assay use. It is supplied as a high-purity solid and is offered in multiple pack sizes, including a 50 mg vial. Specification details and a certificate of analysis are provided for analytical applications.

  • Analytical/reference standard suitable for assay and research applications.
  • Selective cholecystokinin (CCK) receptor antagonist.
  • CAS 103420-77-5.
  • Molecular formula C25H20N4O2; molecular weight 408.45.
  • Purity ≥98.0% (HPLC).
  • Available in multiple pack sizes, including 50 mg.
  • Certificate of analysis and storage instructions provided.

Catalog No. 50-002-43395 Supplier Medchemexpress LLC Supplier No. HY10630150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Devazepide is an analytical reference standard of a selective cholecystokinin (CCK) receptor antagonist intended for research and assay use. It is supplied as a high-purity solid and is offered in multiple pack sizes, including a 50 mg vial. Specification details and a certificate of analysis are provided for analytical applications.

  • Analytical/reference standard suitable for assay and research applications.
  • Selective cholecystokinin (CCK) receptor antagonist.
  • CAS 103420-77-5.
  • Molecular formula C25H20N4O2; molecular weight 408.45.
  • Purity ≥98.0% (HPLC).
  • Available in multiple pack sizes, including 50 mg.
  • Certificate of analysis and storage instructions provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.