Learn More
Medchemexpress LLC Devazepide | 103420-77-5 | MFCD00864500 | 99.6% | 408.45 | C25H20N4O2 | 100 MG
SDP
Catalog No. 5000241884
Encompass_Preferred
Devazepide is a selective, nonpeptide cholecystokinin (CCK) receptor antagonist used in pharmacological research to probe CCK-mediated pathways such as appetite regulation and gastrointestinal motility. It is supplied as a high-purity solid suitable for both in vitro and in vivo studies.
  • Selective CCK1 (CCK-A) receptor antagonist
  • Nonpeptide, orally active research compound
  • High purity (99.55%) suitable for analytical work
  • Molecular weight 408.45 g/mol for dosing calculations
  • Available in solid and solution formats, multiple pack sizes
Catalog No. 50-002-41884 Supplier Medchemexpress LLC Supplier No. HY106301100MG
Devazepide is a selective, nonpeptide cholecystokinin (CCK) receptor antagonist used in pharmacological research to probe CCK-mediated pathways such as appetite regulation and gastrointestinal motility. It is supplied as a high-purity solid suitable for both in vitro and in vivo studies.
  • Selective CCK1 (CCK-A) receptor antagonist
  • Nonpeptide, orally active research compound
  • High purity (99.55%) suitable for analytical work
  • Molecular weight 408.45 g/mol for dosing calculations
  • Available in solid and solution formats, multiple pack sizes