Learn More
RS Hughes Inc DEVCON 5 MIN EPOXY 25ML
SDP
Catalog No. NC9549689
Encompass_Preferred
Devcon 5 Minute Amber Two-Part Epoxy Adhesive - Amber, Base & Accelerator (B/A) - 25 ml Tube - Shore Hardness 85 Shore D, Shear Strength 1900 psi
Catalog No. NC9549689 Supplier RS Hughes Inc Supplier No. 07814314250
Devcon 5 Minute Amber Two-Part Epoxy Adhesive - Amber, Base & Accelerator (B/A) - 25 ml Tube - Shore Hardness 85 Shore D, Shear Strength 1900 psi