Learn More
RS Hughes Inc Devcon 5 minute Amber Two-part Epoxy Adhesive - Base & Accelerator (B/A) - 50mL Cartridge
SDP
Supplier:  RS Hughes Inc VB2 07814314270
Encompass_Preferred
Devcon 5 Minute amber two-part epoxy adhesive is compatible with fabric, concrete, metal and wood materials with a 12 hr cure time. Provides a 3 to 6 min working time. Works in a mix ratio of 1:1. Delivers great performance with a shear strength of 1900 psi and peel strength of 2 to 3 pli. Minimum to maximum operating temperatures are -40 F to +200 F. Comes in a 50 ml Cartridge. This epoxy adhesive is packaged 50 ml per inner.
Catalog No. NC1234528