Learn More
Electron Microscopy Sciences EMS Developer D-19 Powder Replacement, Makes 1 gallon
SDP
Supplier:  Electron Microscopy Sciences 74200
Encompass_Preferred
A high contrast developer for EM films. Available in one gallon (3.8L) or 1L kits. Kits contains Distilled Water (48 degree C/125 degree F), Metol, Sodium Sulfate, Hydroquinone, Sodium Carbonate (monohydrate), Potassium Bromide.
Capacity: Makes 1 gallon
Catalog No. 50-267-66