Learn More
Hach Company Formaldehyde Developing Solution, LR, 500 mL
Supplier:  Hach Company 2257249
Encompass
Reagent for low range formaldehyde determination by the MBTH photometric method. Hach Method 8110 500 mL bottle.
Catalog No. 50-199-5650