Learn More
SKC Inc DEVELOPING SOLUTION, ISOCYANAT
SDP
Catalog No. NC1739792
Encompass
Developing Solution, Isocyanate DeveloperThe Cleaning/Developing Solution is necessary in order to increase the sampling efficiency of both the Surface and Skin SWYPE detection systems. When testing for contamination, the surface is lightly sprayed with the Cleaning/Developing Solution. The solution dissolves the contaminant prior to using the Surface or Skin SWYPE. The SWYPE is designed to wick the solution into the pad, increasing the detection level, particularly on rough or porous surfaces. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC1739792 Supplier SKC Inc Supplier No. 7691062
Developing Solution, Isocyanate DeveloperThe Cleaning/Developing Solution is necessary in order to increase the sampling efficiency of both the Surface and Skin SWYPE detection systems. When testing for contamination, the surface is lightly sprayed with the Cleaning/Developing Solution. The solution dissolves the contaminant prior to using the Surface or Skin SWYPE. The SWYPE is designed to wick the solution into the pad, increasing the detection level, particularly on rough or porous surfaces. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”