Learn More
Electron Microscopy Sciences Developing Tray, Size 11 x 14"
SDP
Catalog No. 5025331
Encompass_Preferred
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 11 x 14"
Catalog No. 50-253-31 Supplier Electron Microscopy Sciences Supplier No. 74361
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 11 x 14"