Learn More
Electron Microscopy Sciences Developing Tray, Size 16 x 20"
SDP
Catalog No. 5025332
Encompass_Preferred
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 16 x 20"
Catalog No. 50-253-32 Supplier Electron Microscopy Sciences Supplier No. 74362
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 16 x 20"