Learn More
Electron Microscopy Sciences Developing Tray, Size 8 x 10"
SDP
Catalog No. 5025330
Encompass_Preferred
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 8 x 10"
Catalog No. 50-253-30 Supplier Electron Microscopy Sciences Supplier No. 74360
Laminated styrene developing trays. Wide cross bars and perimeter ribbing provide rigidity and easy cleaning. 3" (76 mm) deep.
Dimensions: 8 x 10"