Learn More
Meso Scale Discovery U-PLEX Development Pack, 3-plex, 5 plates, 96-well SECTOR Plate, Electrochemiluminescence (ECL), For Assay Development, Immunogenicity, Storage 2-8 °C
SDP
Catalog No. NC2580687
Encompass
U-PLEX Development Pack, 3-plex, 5 plates, 96-well SECTOR Plate, Electrochemiluminescence (ECL), For Assay Development, Immunogenicity, Storage 2-8 °C
Catalog No. NC2580687 Supplier Meso Scale Discovery Supplier No. K15228N2
U-PLEX Development Pack, 3-plex, 5 plates, 96-well SECTOR Plate, Electrochemiluminescence (ECL), For Assay Development, Immunogenicity, Storage 2-8 °C