Learn More
REPLIGEN CORPORATION Repligen XCell ATF 2 Device, High Cell Viability, Gentle Diaphragm Pumping, 1 unit
SDP
Catalog No. NC3345067
Encompass
The Repligen XCell ATF 2 Single-use Device is a high-performance cell retention solution that supports high cell density and viability in bioprocessing. It employs Alternating Tangential Flow (ATF) technology to minimize shear stress, enhancing cell performance. This ready-to-use device is scalable for processes up to 5000 L, featuring integrated tube sets and aseptic connectors that allow seamless transfers to larger systems, differentiating it with its ease of use and effective design.
Catalog No. NC3345067 Supplier REPLIGEN CORPORATION Supplier No. SUATF2S02PESAQ
The Repligen XCell ATF 2 Single-use Device is a high-performance cell retention solution that supports high cell density and viability in bioprocessing. It employs Alternating Tangential Flow (ATF) technology to minimize shear stress, enhancing cell performance. This ready-to-use device is scalable for processes up to 5000 L, featuring integrated tube sets and aseptic connectors that allow seamless transfers to larger systems, differentiating it with its ease of use and effective design.