Learn More
Med Vet International Foley Catheter / Line Securement Device Grip-Lok Small, 3 Inch Length, Sterile, 3200S, 100/Box
Catalog No. 503043995
Encompass_Preferred
Universal Tubing includes Peripherally Inserted Central Catheters (PICC), Central Venous Catheters, IV Lines, Foley Catheters, Feeding Tubes, Surgical Drains, Chest Tubes, Bedside Emergency Equipment or Any place that catheters, tubes or line needs to be secured Made with medical grade, hypoallergenic adhesive that promotes skin integrity Hook-and-loop design allows for repositioning and access to the catheter after securement Low-profile design keeps the device close to the skin while promoting comfort
Catalog No. 50-304-3995 Supplier Med Vet International Supplier No. 1003991BX
Universal Tubing includes Peripherally Inserted Central Catheters (PICC), Central Venous Catheters, IV Lines, Foley Catheters, Feeding Tubes, Surgical Drains, Chest Tubes, Bedside Emergency Equipment or Any place that catheters, tubes or line needs to be secured Made with medical grade, hypoallergenic adhesive that promotes skin integrity Hook-and-loop design allows for repositioning and access to the catheter after securement Low-profile design keeps the device close to the skin while promoting comfort