Learn More
Med Vet International McKesson Grip-Lok Premium Catheter Securement Devices, 6.25 Inch, 18-3400LFC, Each
Catalog No. 503043210
Encompass_Preferred
Catheter Stabilization Device McKesson Premium Large, 6.25 Inch, Sterile Hook-and-loop tab allows for repositioning and access to catheter after securement Low profile with no pins or hard plastic parts Developed with medical-grade, hypoallergenic adhesive to secure device to patient skin Large and glove-friendly peel-away liners for easy application
Catalog No. 50-304-3210 Supplier Med Vet International Supplier No. 1146461EA
Catheter Stabilization Device McKesson Premium Large, 6.25 Inch, Sterile Hook-and-loop tab allows for repositioning and access to catheter after securement Low profile with no pins or hard plastic parts Developed with medical-grade, hypoallergenic adhesive to secure device to patient skin Large and glove-friendly peel-away liners for easy application