Learn More
Medchemexpress LLC HY-15453 100mg , Devimistat CAS:95809-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15453100MG
Encompass_Preferred
HY-15453 100mg Devimistat CAS: 95809-78-2 Devimistat (CPI-613) is a mitochondrial metabolism inhibitor. Devimistat is a lipoic acid antagonist that abrogates mitochondrial energy metabolism to induce apoptosis in various cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9256