Learn More
Medchemexpress LLC HY-15453 10mg Medchemexpress, Devimistat CAS:95809-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1545310MG
Encompass_Preferred
Medchemexpress, HY-15453 10mg Devimistat CAS:95809-78-2 Devimistat (CPI-613) is a lipoic acid analog that inhibits pyruvate dehydrogenase (PDH) and α-ketoglutarate dehydrogenase, disrupts mitochondrial metabolism and shows strong antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9126