Learn More
Medchemexpress LLC Devimistat-d10 | 2586055-61-8 | C22H18D10O2S2 | 100 MG
SDP
Catalog No. 5000376586
Encompass_Preferred
Devimistat-d10 is the deuterium-labeled Devimistat. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled Devimistat
  • For research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-76586 Supplier Medchemexpress LLC Supplier No. HY15453S100MG
Devimistat-d10 is the deuterium-labeled Devimistat. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled Devimistat
  • For research use only
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS