Learn More
Medchemexpress LLC Devimistat-d10 | 2586055-61-8 | C22H18D10O2S2 | 5 MG
SDP
Catalog No. 5000376808
Encompass_Preferred
Devimistat-d10 is the deuterium-labeled Devimistat. It is a white to off-white solid with a purity of 99.79% and a molecular weight of 398.65.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-76808 Supplier Medchemexpress LLC Supplier No. HY15453S5MG
Devimistat-d10 is the deuterium-labeled Devimistat. It is a white to off-white solid with a purity of 99.79% and a molecular weight of 398.65.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.