Learn More
Medchemexpress LLC Devimistat-d10 | 2586055-61-8 | C22H18D10O2S2 | 50 MG
SDP
Catalog No. 5000376883
Encompass_Preferred
Devimistat-d10 is the deuterium-labeled version of Devimistat, intended for research use only and not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-76883 Supplier Medchemexpress LLC Supplier No. HY15453S50MG
Devimistat-d10 is the deuterium-labeled version of Devimistat, intended for research use only and not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.