Learn More
Medchemexpress LLC Devimistat-d10 (CPI-613-d10) | 2586055-61-8 | 99.8% | 398.65 | C22H18D10O2S2 | 10 MG
SDP
Catalog No. 5000286151
Encompass_Preferred
Devimistat-d10 is the deuterium-labeled form of devimistat designed for use as a tracer and as an internal standard in quantitative analytical assays. It is supplied as a high-purity solid suitable for NMR, GC-MS, and LC-MS applications, and includes storage recommendations for both powder and solution states.
  • Can be used as a tracer for metabolic and pharmacokinetic studies.
  • Serves as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity supports accurate and reproducible analytical results.
  • Molecular weight and formula provided for analytical reference.
  • Supplied as a white to off-white solid with specified storage conditions.
  • Stable under recommended storage for reliable long-term use.
Catalog No. 50-002-86151 Supplier Medchemexpress LLC Supplier No. HY15453S10MG
Devimistat-d10 is the deuterium-labeled form of devimistat designed for use as a tracer and as an internal standard in quantitative analytical assays. It is supplied as a high-purity solid suitable for NMR, GC-MS, and LC-MS applications, and includes storage recommendations for both powder and solution states.
  • Can be used as a tracer for metabolic and pharmacokinetic studies.
  • Serves as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity supports accurate and reproducible analytical results.
  • Molecular weight and formula provided for analytical reference.
  • Supplied as a white to off-white solid with specified storage conditions.
  • Stable under recommended storage for reliable long-term use.