Learn More
MSC DeWALT Mobile Workstation, 52 in. Wide, 8-Drawer, Steel
Catalog No. 5000516818
Encompass
This mobile workstation provides a 52-inch wide, 8-drawer design for secure and organized tool storage. It features full-extension soft-close ball-bearing drawer slides, 18/20 gauge steel construction, and 6 in. x 2 in. over-toe locking casters with a 2,000 lbs capacity.
  • Full-extension, soft-close drawer slides support up to 150 lbs.
  • Internal keyed locking system secures tools.
  • Mobile design with a full-grip tubular side handle.
  • 6 in. x 2 in. over-toe locking casters support up to 2,000 lbs.
  • Pre-cut top mat and drawer liners prevent dents.
  • Strong 18/20 gauge steel construction.
  • Includes metal rails, power tool holsters, and bins for custom organization.
  • Made in the USA with global materials.
Catalog No. 50-005-16818 Supplier MSC Supplier No. 06491922
This mobile workstation provides a 52-inch wide, 8-drawer design for secure and organized tool storage. It features full-extension soft-close ball-bearing drawer slides, 18/20 gauge steel construction, and 6 in. x 2 in. over-toe locking casters with a 2,000 lbs capacity.
  • Full-extension, soft-close drawer slides support up to 150 lbs.
  • Internal keyed locking system secures tools.
  • Mobile design with a full-grip tubular side handle.
  • 6 in. x 2 in. over-toe locking casters support up to 2,000 lbs.
  • Pre-cut top mat and drawer liners prevent dents.
  • Strong 18/20 gauge steel construction.
  • Includes metal rails, power tool holsters, and bins for custom organization.
  • Made in the USA with global materials.