Learn More
MSC DEWALT Tool Storage Case, Flat Top, Stackable, Plastic
Catalog No. 5000534962
Encompass
This rugged and durable tool storage case is designed to accommodate cordless and corded power tools, accessories, and attachments. It offers a professional alternative to standard tool boxes and is part of a versatile storage system that allows units to stack and attach for organized storage.
  • Flexible platform allows different combinations.
  • Units stack on top of each other, connected with durable side latches.
  • Bi-material handle on top of each unit for easy and comfortable lifting.
  • Heavy-duty metal latches provide rust resistance.
  • Removable covered cups for organization of small parts and accessories.
  • Removable dividers for organization of drills and bits.
  • Ball bearing slides are reliable for heavy-duty loads.
Catalog No. 50-005-34962 Supplier MSC Supplier No. 52789948
This rugged and durable tool storage case is designed to accommodate cordless and corded power tools, accessories, and attachments. It offers a professional alternative to standard tool boxes and is part of a versatile storage system that allows units to stack and attach for organized storage.
  • Flexible platform allows different combinations.
  • Units stack on top of each other, connected with durable side latches.
  • Bi-material handle on top of each unit for easy and comfortable lifting.
  • Heavy-duty metal latches provide rust resistance.
  • Removable covered cups for organization of small parts and accessories.
  • Removable dividers for organization of drills and bits.
  • Ball bearing slides are reliable for heavy-duty loads.