Learn More
MSC DeWALT Tool Adapter, 35 mm, Plastic, ESD Safe
Catalog No. 502837112
Encompass
This tool adapter connects to dust ports for effective dust collection in concrete, drywall, or woodworking applications. It is designed to fit 35 mm (1.37 inch) OD ports and is compatible with DWV012 and DWV9000 systems. The adapter also features ESD safety.
  • Fits with universal connector for quick and easy connection to tool dust port.
  • Fits 35 mm (1.37 inch) OD ports.
  • Designed for dust collection with tools or shrouds in concrete, drywall, or woodworking applications.
  • ESD safe.
  • Made of plastic.
Catalog No. 50-283-7112 Supplier MSC Supplier No. 53530630
This tool adapter connects to dust ports for effective dust collection in concrete, drywall, or woodworking applications. It is designed to fit 35 mm (1.37 inch) OD ports and is compatible with DWV012 and DWV9000 systems. The adapter also features ESD safety.
  • Fits with universal connector for quick and easy connection to tool dust port.
  • Fits 35 mm (1.37 inch) OD ports.
  • Designed for dust collection with tools or shrouds in concrete, drywall, or woodworking applications.
  • ESD safe.
  • Made of plastic.