Learn More
Taylor Wharton DEWAR 35 LITER
Catalog No. NC9959428
Encompass
35LD cryogenic Dewar designed for storing and dispensing small amounts of liquid nitrogen. Optional equipment includes liquid withdrawal device, dippers and roller base.
Catalog No. NC9959428 Supplier Taylor Wharton Supplier No. 35LDB
35LD cryogenic Dewar designed for storing and dispensing small amounts of liquid nitrogen. Optional equipment includes liquid withdrawal device, dippers and roller base.