Learn More
Electron Microscopy Sciences Dewar Caddie for Lab Grade Dewar
SDP
Catalog No. 501930915
Encompass
The lightweight and sturdy aluminum caddies are ideal for local carrying or decanting lab grade spherical dewars. They can be used repeatedly by simply removing bottom retainer, inserting a new dewar, and replacing the retainer. Gaskets help center dewar and hold it firmly in place. The aluminum top snaps into place.
Use: 61888-11
Catalog No. 50-193-0915 Supplier Electron Microscopy Sciences Supplier No. 6189001
The lightweight and sturdy aluminum caddies are ideal for local carrying or decanting lab grade spherical dewars. They can be used repeatedly by simply removing bottom retainer, inserting a new dewar, and replacing the retainer. Gaskets help center dewar and hold it firmly in place. The aluminum top snaps into place.
Use: 61888-11