Learn More
Electron Microscopy Sciences Dewar Caddie for Lab Grade Dewar
SDP
Supplier:  Electron Microscopy Sciences 6189002
Encompass
The lightweight and sturdy aluminum caddies are ideal for local carrying or decanting lab grade spherical dewars. They can be used repeatedly by simply removing bottom retainer, inserting a new dewar, and replacing the retainer. Gaskets help center dewar and hold it firmly in place. The aluminum top snaps into place.
Use: 61888-12
Catalog No. 50-193-0916