Learn More
Chemglass Life Sciences Condenser, Dewar Type, 24/40 Joint, 250 x 50mm
SDP
Catalog No. CG1209A27
Encompass_Preferred
Condenser, Dewar Type, 24/40 Joint, 250 x 50mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.
Catalog No. CG1209A27 Supplier Chemglass Life Sciences Supplier No. CG1209A27
Condenser, Dewar Type, 24/40 Joint, 250 x 50mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.