Learn More
Ace Glass, Inc. Dewar flask, 1000ml, Cylindrical form
SDP
Catalog No. NC0537047
Encompass
1000ml Cylindrical form, silvered and evacuated. Furnished with an aluminum base and plastic mesh. Intended for use with liquid nitrogen only. Performance is not guaranteed with dry ice, acetone or other solvents.
Catalog No. NC0537047 Supplier Ace Glass, Inc. Supplier No. 707515
1000ml Cylindrical form, silvered and evacuated. Furnished with an aluminum base and plastic mesh. Intended for use with liquid nitrogen only. Performance is not guaranteed with dry ice, acetone or other solvents.