Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 4300mL Capacity, 305mm ID X 143mm Inside Depth, 178mm OD X 365mm Height
SDP
Catalog No. CG159503
Encompass_Preferred
Aluminum Caddies for Spherical Dewar Flask, Fits 2L. Affords protection during transportation or pouring of CG-1596, spherical dewar flasks. Caddies can be used over and over again by simply removing the bottom retainer, inserting another dewar, and replacing the retainer. Gaskets help center the dewar and hold it firmly in place.
Catalog No. CG159503 Supplier Chemglass Life Sciences Supplier No. CG159503
Aluminum Caddies for Spherical Dewar Flask, Fits 2L. Affords protection during transportation or pouring of CG-1596, spherical dewar flasks. Caddies can be used over and over again by simply removing the bottom retainer, inserting another dewar, and replacing the retainer. Gaskets help center the dewar and hold it firmly in place.