Learn More
Cole-Parmer Dilvac Universal-Style Stainless Steel and Glass Dewar Flask with Handle and Lid, 1 L
Catalog No. NC1504677 Shop All Cole-Parmer Products
Encompass_Preferred
Universal-Style Stainless Steel and Glass Dewar Flask with Handle and Lid, 1 L, Toggle clamps secure lid for safe transportationStainless steel containersHygienic and won''t rust or corrodeBorosilicate glass inner vesselStainless steel lid with self-contained insulationBuilt-in handle for easy handling
Catalog No. NC1504677 Supplier Cole-Parmer Supplier No. 0377413
Universal-Style Stainless Steel and Glass Dewar Flask with Handle and Lid, 1 L, Toggle clamps secure lid for safe transportationStainless steel containersHygienic and won''t rust or corrodeBorosilicate glass inner vesselStainless steel lid with self-contained insulationBuilt-in handle for easy handling