Learn More
Mitegen 3 Pack Foam Dewar Value Bundle, Features 500mL, 800mL, 1400mL Vessels, Each Is Lightweight, Shatterproof, For Lab Chilling & Cryogenic Use
SDP
Supplier:  Mitegen MFDBD2
Encompass
This Foam Dewar Value Bundle provides a versatile and safe solution for a wide range of laboratory applications, offering three essential sizes for precise temperature maintenance. The 3-pack includes one each of 500 mL (Small), 800 mL (Standard), and 1400 mL (Large) foam dewars. Crafted from robust, high-density foam, all dewars in this bundle provide excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of sensitive samples or reagents. Their lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Each dewar features a wide, accessible opening and a stable base, making them reliable and indispensable additions for quick chilling, temporary cryogenic storage, and other temperature-sensitive processes across various laboratory settings.
Catalog No. NC1866261