Learn More
Mitegen 4 Pack Foam Dewar Bundle, Features 500mL, 800mL, 1400mL, 1800mL Vessels, Each Is Lightweight, Shatterproof, For Lab Chilling & Cryogenic Use
SDP
Catalog No. NC1866260
Encompass
This Foam Dewar Bundle provides a versatile and safe solution for a wide range of laboratory applications, offering four essential sizes for precise temperature maintenance. The 4-pack includes one each of 500 mL (Small), 800 mL (Standard), 1400 mL (Large), and 1800 mL (Tall) foam dewars. Crafted from robust, high-density foam, all dewars in this bundle provide excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of sensitive samples or reagents. Their lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Each dewar features a wide, accessible opening and a stable base, making them reliable and indispensable additions for quick chilling, temporary cryogenic storage, and other temperature-sensitive processes across various laboratory settings.
Catalog No. NC1866260 Supplier Mitegen Supplier No. MFDBD1
This Foam Dewar Bundle provides a versatile and safe solution for a wide range of laboratory applications, offering four essential sizes for precise temperature maintenance. The 4-pack includes one each of 500 mL (Small), 800 mL (Standard), 1400 mL (Large), and 1800 mL (Tall) foam dewars. Crafted from robust, high-density foam, all dewars in this bundle provide excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of sensitive samples or reagents. Their lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Each dewar features a wide, accessible opening and a stable base, making them reliable and indispensable additions for quick chilling, temporary cryogenic storage, and other temperature-sensitive processes across various laboratory settings.