Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height
SDP
Catalog No. CG159403
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159403 Supplier Chemglass Life Sciences Supplier No. CG159403
Dewar Flask, Wide Mouth, Metal Housing, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.