Learn More
Chemglass Life Sciences Vacuum Trap, Dewar Type, PTFE Stopcock, 1000mL Reservoir, Inner Chamber ID: 85mm, Inner Chamber Depth: 300mm, 650mm OAH x 120mm Width
SDP
Catalog No. CG451702
Encompass_Preferred
Vacuum Trap, Dewar Type, PTFE Stopcock, 1000mL Reservoir, Inner Chamber ID: 85mm, Inner Chamber Depth: 300mm, 650mm OAH x 120mm Width. Dry ice, Dewar style trap condenser designed for large volume solvent removal. Lower reservoir has a capacity of 1000mL, with a 4mm PTFE stopcock. Serrated hose connections are 12mm at the smallest serration and 14mm at the largest serration. Optional UHMW Lid prevents evaporation of dry ice/acetone in trap.PLEASE NOTE: CG-4517 TRAPS ARE AVAILABLE PLASTIC COATED. PLEASE CONTACT US FOR DETAILS.
Catalog No. CG451702 Supplier Chemglass Life Sciences Supplier No. CG451702
Vacuum Trap, Dewar Type, PTFE Stopcock, 1000mL Reservoir, Inner Chamber ID: 85mm, Inner Chamber Depth: 300mm, 650mm OAH x 120mm Width. Dry ice, Dewar style trap condenser designed for large volume solvent removal. Lower reservoir has a capacity of 1000mL, with a 4mm PTFE stopcock. Serrated hose connections are 12mm at the smallest serration and 14mm at the largest serration. Optional UHMW Lid prevents evaporation of dry ice/acetone in trap.PLEASE NOTE: CG-4517 TRAPS ARE AVAILABLE PLASTIC COATED. PLEASE CONTACT US FOR DETAILS.