Learn More
Biogenex Laboratories EZ-DeWax (Manual/i6000*)
SDP
Catalog No. NC9866306
Encompass_Preferred
BioGenex is the inventor of Antigen Retrieval enabling technology. Antigen Retrieval is an effective way of unmasking antigenic epitopes on the surface of formalin-fixed, parraffin-embedded tissue sections using microwave heating. Covered by exclusive patents issued to BioGenex, this method to recover antigenicity in formalin-fixed, paraffin-embedded tissue sections has been routinely practiced in laboratories throughout the world.The Antigen Retrieval technique breaks the formalin induced cross-linking bonds between epitopes and unrelated proteins, thereby allowing better penetration of antibody and accessibility of epitopes.
Catalog No. NC9866306 Supplier Biogenex Laboratories Supplier No. HK5855K
BioGenex is the inventor of Antigen Retrieval enabling technology. Antigen Retrieval is an effective way of unmasking antigenic epitopes on the surface of formalin-fixed, parraffin-embedded tissue sections using microwave heating. Covered by exclusive patents issued to BioGenex, this method to recover antigenicity in formalin-fixed, paraffin-embedded tissue sections has been routinely practiced in laboratories throughout the world.The Antigen Retrieval technique breaks the formalin induced cross-linking bonds between epitopes and unrelated proteins, thereby allowing better penetration of antibody and accessibility of epitopes.