Learn More
Epredia™ Dewax and HIER Buffer L,M,H
Catalog No. TA999DHBM Shop All Epredia Products
Change view
Click to view available options
Solution Type:
Buffer - High
Buffer - Low
Buffer - Medium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Solution Type
TA999DHBM Buffer - Medium
TA999DHBH Buffer - High
TA999DHBL Buffer - Low
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TA999DHBM Supplier Epredia™ Supplier No. TA999DHBM
Add to Cart
Add to Cart
Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.
Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.
Specifications
Product Type Dewax and HIER Buffer
Quantity 10 x 100 mL
Solution Type Buffer - Medium